Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Difluoroethyl Acetate 98.0+%, TCI America™
SDP

Catalog No. D50405G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D50405G 5 g
D504025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D50405G Supplier TCI America Supplier No. D50405G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1550-44-3
Molecular Formula C4H6F2O2
Molecular Weight (g/mol) 124.087
MDL Number MFCD00069133
InChI Key PFJLHSIZFYNAHH-UHFFFAOYSA-N
Synonym Acetatic Acid 2,2-Difluoroethyl Ester
PubChem CID 137074
IUPAC Name 2,2-difluoroethyl acetate
SMILES CC(=O)OCC(F)F

Specifications

Color Yellow
Boiling Point 106°C
Quantity 5 g
UN Number 3272
Formula Weight 124.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2-Difluoroethyl Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.