accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3S

CAS RN 1551-27-5

IUPAC Name: 2-propylthiophene
Synonyms: thiophene, 2-propyl-
Molecular Weight (g/mol): 126.22
Molecular Formula: C7H10S
InChi Key: BTXIJTYYMLCUHI-UHFFFAOYSA-N
SMILES: CCCC1=CC=CS1
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)