Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-n-Propylthiophene, 97%, Thermo Scientific™

Catalog No. AAL0305206
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0305206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0305206 Supplier Thermo Scientific Chemicals Supplier No. L0305206

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1551-27-5
Molecular Formula C7H10S
Molecular Weight (g/mol) 126.217
MDL Number MFCD00014533
InChI Key BTXIJTYYMLCUHI-UHFFFAOYSA-N
Synonym 2-n-propylthiophene, thiophene, 2-propyl, isopropylthiophene, 2-n-propyl thiophene, 2-propyl-thiophene, 2-propyl thiophene, thiophene, propyl, thiophene,2-propyl, 2-propylthiophene, 4-hydroxy-benzenesulfonamide
PubChem CID 73771
IUPAC Name 2-propylthiophene
SMILES CCCC1=CC=CS1

Specifications

Density 0.984
Boiling Point 158°C to 160°C
Flash Point 32°C (90°F)
Odor Characteristic
Refractive Index 1.505
Quantity 5 g
UN Number UN1993
Beilstein 105753
Formula Weight 126.22
Percent Purity 97%
Chemical Name or Material 2-n-Propylthiophene
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 216-288-8
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.