accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONNSNO

CAS RN 155106-73-3

IUPAC Name: 2-{4-[(2H-1,3-benzodioxol-5-yl)methyl]piperazin-1-yl}-1,3-benzothiazole
Synonyms: 2-[4-(2H-1,3-benzodioxol-5-ylmethyl)piperazin-1-yl]-1,3-benzothiazole
Molecular Weight (g/mol): 353.44
Molecular Formula: C19H19N3O2S
InChi Key: BYHKGNWKJMGHGE-UHFFFAOYSA-N
SMILES: C(N1CCN(CC1)C1=NC2=CC=CC=C2S1)C1=CC=C2OCOC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results