accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CNNOO

CAS RN 15513-52-7

IUPAC Name: 2,6-dimethyl-3-nitropyridine
Synonyms: 2,6-dimethyl-3-nitropyridine
Molecular Weight (g/mol): 152.15
Molecular Formula: C7H8N2O2
InChi Key: AETHUDGJSSKZKT-UHFFFAOYSA-N
SMILES: CC1=CC=C(C(C)=N1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results