accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHNOOOCH3OOH3COOCH3OOCH3SOOCH3

CAS RN 155155-64-9

IUPAC Name: methyl 4-(acetyloxy)-5-acetamido-2-(phenylsulfanyl)-6-[1,2,3-tris(acetyloxy)propyl]oxane-2-carboxylate
Synonyms: methyl 4-(acetyloxy)-5-acetamido-2-(phenylsulfanyl)-6-[1,2,3-tris(acetyloxy)propyl]oxane-2-carboxylate
Molecular Weight (g/mol): 583.61
Molecular Formula: C26H33NO12S
InChi Key: NPLKOUSSDBQHEN-UHFFFAOYNA-N
SMILES: COC(=O)C1(CC(OC(C)=O)C(NC(C)=O)C(O1)C(OC(C)=O)C(COC(C)=O)OC(C)=O)SC1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results