Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neu5Ac[1Me,4789Ac]-SPh 97.0+%, TCI America™
SDP

Catalog No. M17061G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M17061G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M17061G Supplier TCI America Supplier No. M17061G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 155155-64-9
Molecular Formula C26H33NO12S
Molecular Weight (g/mol) 583.605
InChI Key NPLKOUSSDBQHEN-MVWXVPEESA-N
Synonym Methyl (Phenyl 5-Acetamido-4,7,8,9-tetra-O-acetyl-3,5-dideoxy-2-thio-D-glycero-D-galacto-2-nonulopyranosid)onate, Neu5Ac[1Me,4789Ac]-SPh
PubChem CID 11734585
IUPAC Name methyl (4S,5R,6R)-5-acetamido-4-acetyloxy-2-phenylsulfanyl-6-[(1S,2R)-1,2,3-triacetyloxypropyl]oxane-2-carboxylate
SMILES CC(=O)NC1C(CC(OC1C(C(COC(=O)C)OC(=O)C)OC(=O)C)(C(=O)OC)SC2=CC=CC=C2)OC(=O)C

Specifications

Melting Point 183°C
Color White
Quantity 1 g
Formula Weight 583.61
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 5-Acetamido-4,7,8,9-tetra-O-acetyl-3,5-dideoxy-2-S-phenyl-2-thio-D-glycero-D-galacto-2-nonulo
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.