accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15525-15-2

IUPAC Name: tetraphenylboranuide; tetraphenylphosphanium
Synonyms: tetraphenylphosphonium tetraphenylborate
Molecular Weight (g/mol): 658.63
Molecular Formula: C48H40BP
InChi Key: QYFWPOPFFBCTLK-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)[B-](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results