Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraphenylphosphonium Tetraphenylborate 98.0+%, TCI America™
SDP

Catalog No. T132925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T132925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T132925G Supplier TCI America Supplier No. T132925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 15525-15-2
Molecular Formula C48H40BP
Molecular Weight (g/mol) 658.632
MDL Number MFCD00145389
InChI Key QYFWPOPFFBCTLK-UHFFFAOYSA-N
PubChem CID 9831062
IUPAC Name tetraphenylboranuide;tetraphenylphosphanium
SMILES [B-](C1=CC=CC=C1)(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4.C1=CC=C(C=C1)[P+](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4

Specifications

Melting Point 306°C
Color White-Yellow
Quantity 25 g
Formula Weight 658.63
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Tetraphenylphosphonium Tetraphenylborate
EINECSNumber (3)-3389
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.