accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15525-45-8

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 248.22
Molecular Formula: Cl4Pd
InChi Key: BRXVSUHTXBZCMN-UHFFFAOYSA-N
SMILES: [Cl].[Cl].[Cl].[Cl].[Pd++]
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results