accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2FFFI

CAS RN 155403-06-8

IUPAC Name: 4-iodo-3-(trifluoromethyl)aniline
Synonyms: 4-iodo-3-(trifluoromethyl)aniline
Molecular Weight (g/mol): 287.02
Molecular Formula: C7H5F3IN
InChi Key: ZPVVSYSGZZPGPH-UHFFFAOYSA-N
SMILES: NC1=CC(=C(I)C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results