accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHNSOONHOHCl

CAS RN 155558-32-0

IUPAC Name: 5-(1,4-diazepane-1-sulfonyl)-1,2-dihydroisoquinolin-1-one hydrochloride
Synonyms: hydroxyfasudil hydrochloride
Molecular Weight (g/mol): 343.83
Molecular Formula: C14H18ClN3O3S
InChi Key: XWWFOUVDVJGNNG-UHFFFAOYSA-N
SMILES: Cl.O=C1NC=CC2=C(C=CC=C12)S(=O)(=O)N1CCCNCC1
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results