Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Hydroxyfasudil

Catalog No. 3906022MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-060-22MG 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 39-060-22MG Supplier MilliporeSigma™ Supplier No. 3906022MG
Add to Cart
Add to Cart

A cell-permeable, hydroxylated metabolite of HA 1077 that displays anti-anginal properties.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Formulation A 10 mM (2mg/582μl) solution in H2O.
Description 98% by HPLC
Target Proteins Rho-kinase
CAS 155558-32-0
Molecular Formula C14H17N3O3S·HCl
Formula Weight 343.8g/mol
Content And Storage Protect from light
Form Liquid
Quantity 2 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.