accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 155775-31-8

IUPAC Name: trinickel(2+) tetrahydroxide carbonate
Synonyms: nickel carbonate hydroxide
Molecular Weight (g/mol): 304.12
Molecular Formula: CH4Ni3O7
InChi Key: ATYNVYNPWGEMMI-UHFFFAOYSA-H
SMILES: [OH-].[OH-].[OH-].[OH-].[Ni++].[Ni++].[Ni++].[O-]C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)