Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel(II) carbonate (basic) hydrate, Puratronic™, 99.996% (metals basis), Thermo Scientific™

Catalog No. AA1081414
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1081414 25 g
AA1081406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1081414 Supplier Thermo Scientific Chemicals Supplier No. 1081414

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 155775-31-8
Molecular Formula CH4Ni3O7
Molecular Weight (g/mol) 304.12
MDL Number MFCD00016262
InChI Key ATYNVYNPWGEMMI-UHFFFAOYSA-H
Synonym nickel 3+ ;carbonate;hydroxide;hydrate, nickel carbonate, basic hydrate trace metals basis
PubChem CID 86176052
IUPAC Name nickel(3+);carbonate;hydroxide;hydrate
SMILES [OH-].[OH-].[OH-].[OH-].[Ni++].[Ni++].[Ni++].[O-]C([O-])=O

Specifications

Color Green
Physical Form Powder
Quantity 25 g
Linear Formula NiCO3·2Ni(OH)2·xH2O
UN Number UN3077
Solubility Information Insoluble in water.
Formula Weight 304.18 (Anhydrous)
Percent Purity 99.996%
Grade Puratronic
Assay (metals basis)
Chemical Name or Material Nickel(II) carbonate (basic) hydrate
Show More Show Less
Hazard Category H302+H332-H315-H317-H319-H334-H341-H350i-H360D-H372
Hazard Statement GHS H Statement
H334-H350-H360-H372-H341-H302-H332-H315-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P272-P280-P281-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P333+P313-P342+P311-P362-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 240-408-8
RTECSNumber QR6260000
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.