accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 155849-49-3

IUPAC Name: 4-(trifluoromethyl)piperidine hydrochloride
Synonyms: 4-(trifluoromethyl)piperidine hydrochloride
Molecular Weight (g/mol): 189.61
Molecular Formula: C6H11ClF3N
InChi Key: KMUKXPQVPQUCDB-UHFFFAOYSA-N
SMILES: Cl.FC(F)(F)C1CCNCC1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results