CAS RN 156-87-6
3-Amino-1-propanol, 99%
CAS: 156-87-6 Molecular Formula: C3H9NO Molecular Weight (g/mol): 75.111 MDL Number: MFCD00008223 InChI Key: WUGQZFFCHPXWKQ-UHFFFAOYSA-N Synonym: 3-aminopropanol,3-amino-1-propanol,propanolamine,1-propanol, 3-amino,3-propanolamine,1-amino-3-propanol,3-hydroxypropylamine,3-aminopropyl alcohol,1,3-propanolamine,beta-alaninol PubChem CID: 9086 IUPAC Name: 3-aminopropan-1-ol SMILES: C(CN)CO
3-Amino-1-propanol, ≥99.5% (GC), MilliporeSigma™ Supelco™
3-Amino-1-propanol is a linear primary alkanolamine.
3-Amino-1-propanol, 99%
CAS: 156-87-6 MDL Number: MFCD00008223 InChI Key: WUGQZFFCHPXWKQ-UHFFFAOYSA-N Synonym: 3-aminopropanol,3-amino-1-propanol,propanolamine,1-propanol, 3-amino,3-propanolamine,1-amino-3-propanol,3-hydroxypropylamine,3-aminopropyl alcohol,1,3-propanolamine,beta-alaninol PubChem CID: 9086 IUPAC Name: 3-aminopropan-1-ol SMILES: C(CN)CO
3-Amino-1-propanol 99.0+%, TCI America™
CAS: 156-87-6 Molecular Formula: C3H9NO Molecular Weight (g/mol): 75.111 MDL Number: MFCD00008223 InChI Key: WUGQZFFCHPXWKQ-UHFFFAOYSA-N Synonym: 3-aminopropanol,3-amino-1-propanol,propanolamine,1-propanol, 3-amino,3-propanolamine,1-amino-3-propanol,3-hydroxypropylamine,3-aminopropyl alcohol,1,3-propanolamine,beta-alaninol PubChem CID: 9086 IUPAC Name: 3-aminopropan-1-ol SMILES: C(CN)CO
Sigma Aldrich 3-Amino-1-propanol
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich 3-Amino-1-propanol
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
MP Biomedicals, Inc 3-Aminopropanol, MP Biomedicals
CAS: 156-87-6 Molecular Formula: C3H9NO Molecular Weight (g/mol): 75.111 InChI Key: WUGQZFFCHPXWKQ-UHFFFAOYSA-N Synonym: 3-aminopropanol,3-amino-1-propanol,propanolamine,1-propanol, 3-amino,3-propanolamine,1-amino-3-propanol,3-hydroxypropylamine,3-aminopropyl alcohol,1,3-propanolamine,beta-alaninol PubChem CID: 9086 IUPAC Name: 3-aminopropan-1-ol SMILES: C(CN)CO