Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-1-propanol, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019335 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9335 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9335 Supplier MilliporeSigma Supelco Supplier No. 018875MLF
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Amino-1-propanol is a linear primary alkanolamine.

Featured Industries: Environmental
TRUSTED_SUSTAINABILITY

Specifications

CAS 156-87-6
Ignition Point 770°F
Vapor Pressure 2.1 mmHg (20°C)
Vapor Density 2.59 (vs. air)
Density 0.982 g/mL (at 20°C)
Flash Point 100°C - Pensky-Martens closed cup
Linear Formula HO(CH2)3 NH2
MDL Number MFCD00008223
Melting Point 10°C to 12°C
Boiling Point 184°C to 187°C
Synonym 3-Aminopropyl alcohol
Type Analytical Standard
Quantity 5 mL
Formula Weight 75.11
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.