accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 156021-08-8

IUPAC Name: 4-methyl-2,5-diphenylpyridine
Synonyms: 4-methyl-2,5-diphenylpyridine
Molecular Weight (g/mol): 245.33
Molecular Formula: C18H15N
InChi Key: YHQCNNMCDMYPIA-UHFFFAOYSA-N
SMILES: CC1=CC(=NC=C1C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results