Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methyl-2,5-diphenylpyridine 98.0+%, TCI America™
SDP

Catalog No. M28265G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M28265G 5 g
M28261G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M28265G Supplier TCI America Supplier No. M28265G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 156021-08-8
Molecular Formula C18H15N
Molecular Weight (g/mol) 245.33
MDL Number MFCD13152024
InChI Key YHQCNNMCDMYPIA-UHFFFAOYSA-N
PubChem CID 14924986
IUPAC Name 4-methyl-2,5-diphenylpyridine
SMILES CC1=CC(=NC=C1C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 129°C
Color Yellow
Quantity 5 g
Formula Weight 245.33
Percent Purity ≥98.0% (GC)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material 4-Methyl-2,5-diphenylpyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.