accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClIClCs

CAS RN 15605-42-2

IUPAC Name: caesium(1+) dichloroiodanuide
Synonyms: caesium(1+) dichloroiodanuide
Molecular Weight (g/mol): 330.71
Molecular Formula: Cl2CsI
InChi Key: WYPYOYILFBSLTD-UHFFFAOYSA-N
SMILES: [Cs+].Cl[I-]Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results