Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cesium Dichloro Iodide, MP Biomedicals

Catalog No. ICN226156 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN226156 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN226156 Supplier MP Biomedicals, Inc Supplier No. 0522615601

Chemical Identifiers

CAS 15605-42-2
Molecular Formula Cl2CsI
Molecular Weight (g/mol) 330.71
InChI Key WYPYOYILFBSLTD-UHFFFAOYSA-N
Synonym cesium dichloroiodate, caesium 1+ dichloroiodanuide, cesium dichloroiodide, cesium dichloro iodide, caesium 1+ ion dichloroiodanuide, cesium dichloroiodide trace metals basis
PubChem CID 23685551
SMILES Cl[I-]Cl.[Cs+]

Specifications

Quantity 1 g
Chemical Name or Material Cesium Dichloro Iodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.