accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3SOONH3CClONON

CAS RN 156153-43-4

IUPAC Name: 2-{[7-(dimethylsulfamoyl)-2,1,3-benzoxadiazol-4-yl](methyl)amino}acetyl chloride
Synonyms: 2-{[7-(dimethylsulfamoyl)-2,1,3-benzoxadiazol-4-yl](methyl)amino}acetyl chloride
Molecular Weight (g/mol): 332.76
Molecular Formula: C11H13ClN4O4S
InChi Key: NAIIBGUHMHXMNS-UHFFFAOYSA-N
SMILES: CN(C)S(=O)(=O)C1=CC=C(N(C)CC(Cl)=O)C2=NON=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results