Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DBD-COCl 93.0+%, TCI America™
SDP

Catalog No. A5558100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A5558100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A5558100MG Supplier TCI America Supplier No. A5558100MG
Add to Cart
Add to Cart

[for HPLC Labeling]

Chemical Identifiers

CAS 156153-43-4
Molecular Formula C11H13ClN4O4S
Molecular Weight (g/mol) 332.759
MDL Number MFCD00191377
InChI Key NAIIBGUHMHXMNS-UHFFFAOYSA-N
Synonym 4-(N,N-Dimethylaminosulfonyl)-7-(N-chloroformylmethyl-N-methylamino)benzofurazan, 4-(N,N-Dimethylaminosulfonyl)-7-(N-chloroformylmethyl-N-methylamino)-2,1,3-benzoxadiazole
PubChem CID 133003
IUPAC Name 2-[[4-(dimethylsulfamoyl)-2,1,3-benzoxadiazol-7-yl]-methylamino]acetyl chloride
SMILES CN(C)S(=O)(=O)C1=CC=C(C2=NON=C12)N(C)CC(=O)Cl

Specifications

Color Yellow
Quantity 100 mg
UN Number 3261
Formula Weight 332.76
Percent Purity ≥93.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material DBD-COCl [=4-(N,N-Dimethylaminosulfonyl)-7-(N-chloroformylmethyl-N-methylamino)-2,1,3-benzoxa
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.