accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFFFFFFFFFFFFFFFFFFFFFFFFFHO

CAS RN 15622-57-8

IUPAC Name: 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,14-heptacosafluorotetradecan-1-ol
Synonyms: 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,14-heptacosafluorotetradecan-1-ol
Molecular Weight (g/mol): 700.13
Molecular Formula: C14H3F27O
InChi Key: WYCXYEWKMLSDBQ-UHFFFAOYSA-N
SMILES: OCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results