Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1H,1H-Perfluoro-1-tetradecanol, 96%

Catalog No. AAL1660906
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1660906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1660906 Supplier Thermo Scientific Chemicals Supplier No. L1660906

missing translation for 'validMainframeMsg'

CAS: 15622-57-8 | C14H3F27O | 700.134 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15622-57-8
Molecular Formula C14H3F27O
Molecular Weight (g/mol) 700.134
MDL Number MFCD00153264
InChI Key WYCXYEWKMLSDBQ-UHFFFAOYSA-N
Synonym 1h,1h-perfluoro-1-tetradecanol, heptacosafluorotridecyl methanol, 1h,1h-perfluorotetradecan-1-ol
PubChem CID 2776373
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,14-heptacosafluorotetradecan-1-ol
SMILES C(C(C(C(C(C(C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O

Specifications

Melting Point 137°C to 140°C
Boiling Point 130°C (60 mmHg)
Quantity 5 g
Formula Weight 700.14
Percent Purity 96%
Chemical Name or Material 1H,1H-Perfluoro-1-tetradecanol
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.