accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFFFFOSOOFFF

CAS RN 156241-41-7

IUPAC Name: 1,1,1,3,3,3-hexafluoropropan-2-yl trifluoromethanesulfonate
Synonyms: 1,1,1,3,3,3-hexafluoropropan-2-yl trifluoromethanesulfonate
Molecular Weight (g/mol): 300.09
Molecular Formula: C4HF9O3S
InChi Key: NRHQWNHARTXNOE-UHFFFAOYSA-N
SMILES: FC(F)(F)C(OS(=O)(=O)C(F)(F)F)C(F)(F)F
Grade 
  • (1)
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results