accessibility menu, dialog, popup

UserName

Viewing profile as user:
IrOOOOOOH3CCH3CH3H3CH3CH3C

CAS RN 15635-87-7

IUPAC Name: iridium(3+) tris((2Z)-4-oxopent-2-en-2-olate)
Synonyms: iridium(3+) tris((2Z)-4-oxopent-2-en-2-olate)
Molecular Weight (g/mol): 489.54
Molecular Formula: C15H21IrO6
InChi Key: HLYTZTFNIRBLNA-LNTINUHCSA-K
SMILES: [Ir+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)