Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(2,4-pentanedionato)iridium(III), TCI America™
SDP

Catalog No. T25571G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T25571G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T25571G Supplier TCI America Supplier No. T25571G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 15635-87-7
Molecular Formula C15H21IrO6
Molecular Weight (g/mol) 489.54
MDL Number MFCD00015353
InChI Key HLYTZTFNIRBLNA-LNTINUHCSA-K
Synonym Iridium(III) Acetylacetonate, Acetylacetonato Iridium(III) Salt
PubChem CID 117590592
IUPAC Name iridium(3+) tris((2Z)-4-oxopent-2-en-2-olate)
SMILES [Ir+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Melting Point 271°C
Quantity 1 g
Formula Weight 489.54
Physical Form Crystalline Powder
Chemical Name or Material Tris(2,4-pentanedionato)iridium(III)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.