accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OBrFF

CAS RN 156573-09-0

IUPAC Name: 1-bromo-4-ethoxy-2,3-difluorobenzene
Synonyms: 1-bromo-4-ethoxy-2,3-difluorobenzene
Molecular Weight (g/mol): 237.04
Molecular Formula: C8H7BrF2O
InChi Key: OJZVBJOHBGYINN-UHFFFAOYSA-N
SMILES: CCOC1=C(F)C(F)=C(Br)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results