Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-2,3-difluorophenetole 98.0+%, TCI America™
SDP

Catalog No. B38845G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38845G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B38845G Supplier TCI America Supplier No. B38845G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 156573-09-0
Molecular Formula C8H7BrF2O
Molecular Weight (g/mol) 237.044
MDL Number MFCD07368018
InChI Key OJZVBJOHBGYINN-UHFFFAOYSA-N
Synonym 1-Bromo-4-ethoxy-2,3-difluorobenzene
PubChem CID 26985491
IUPAC Name 1-bromo-4-ethoxy-2,3-difluorobenzene
SMILES CCOC1=C(C(=C(C=C1)Br)F)F

Specifications

Melting Point 31°C
Color White
Quantity 5 g
Formula Weight 237.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Bromo-2,3-difluorophenetole
EINECSNumber (3)-4612
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.