accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH2N

CAS RN 15667-63-7

IUPAC Name: 1-cyanoprop-2-en-1-yl acetate
Synonyms: 1-cyanoprop-2-en-1-yl acetate
Molecular Weight (g/mol): 125.13
Molecular Formula: C6H7NO2
InChi Key: MEHLEOUIWVWVBF-UHFFFAOYNA-N
SMILES: CC(=O)OC(C=C)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results