Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Cyano-2-propenyl Acetate 97.0+%, TCI America™
SDP

Catalog No. A11965G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A11965G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. A11965G Supplier TCI America Supplier No. A11965G
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 15667-63-7
Molecular Formula C6H7NO2
Molecular Weight (g/mol) 125.127
MDL Number MFCD00010336
InChI Key MEHLEOUIWVWVBF-UHFFFAOYSA-N
Synonym Acetic Acid 1-Cyano-2-propenyl Ester, DL-2-Acetoxy-3-butenenitrile, 1-Cyanoallyl Acetate
PubChem CID 85891
IUPAC Name 1-cyanoprop-2-enyl acetate
SMILES CC(=O)OC(C=C)C#N

Specifications

Color Yellow
Boiling Point 74°C
Quantity 5 g
UN Number 3276
Formula Weight 125.13
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Cyano-2-propenyl Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.