accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15673-79-7

IUPAC Name: barium(2+) [(2R,3S,4R)-3,4,5-trihydroxyoxolan-2-yl]methyl phosphate
Synonyms: barium(2+) D-ribofuranose 5-phosphate(2-)
Molecular Weight (g/mol): 365.42
Molecular Formula: C5H9BaO8P
InChi Key: VNFDCDZWBYRPMC-QHEAFLAXSA-L
SMILES: [Ba++].OC1O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H]1O
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)