Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ribose-5-phosphate Barium Salt Hydrate 95.0+%, TCI America™
SDP

Catalog No. R00261G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R00261G 1 g
R0026100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R00261G Supplier TCI America Supplier No. R00261G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 15673-79-7
Molecular Formula C5H9BaO8P
Molecular Weight (g/mol) 365.42
MDL Number MFCD00047534
InChI Key VNFDCDZWBYRPMC-QHEAFLAXSA-L
PubChem CID 123134315
IUPAC Name barium(2+) [(2R,3S,4R)-3,4,5-trihydroxyoxolan-2-yl]methyl phosphate
SMILES [Ba++].OC1O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H]1O

Specifications

Color White-Yellow
Quantity 1 g
UN Number 1564
Formula Weight 365.42
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Ribose-5-phosphate Barium Salt Hydrate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.