accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SH3CN

CAS RN 15679-12-6

IUPAC Name: 2-ethyl-4-methyl-1,3-thiazole
Synonyms: 2-ethyl-4-methyl-1,3-thiazole
Molecular Weight (g/mol): 127.21
Molecular Formula: C6H9NS
InChi Key: VGRVKVGGUPOCMT-UHFFFAOYSA-N
SMILES: CCC1=NC(C)=CS1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results