Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethyl-4-methylthiazole 98.0+%, TCI America™
SDP

Catalog No. E04565G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E04565G 5 g
E045625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E04565G Supplier TCI America Supplier No. E04565G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 15679-12-6
Molecular Formula C6H9NS
Molecular Weight (g/mol) 127.205
MDL Number MFCD00036621
InChI Key VGRVKVGGUPOCMT-UHFFFAOYSA-N
Synonym 2-ethyl-4-methylthiazole, 2-ethyl-4-methyl thiazole, thiazole, 2-ethyl-4-methyl, unii-lvc1zv6x4u, 4-methyl-2-ethylthiazole, lvc1zv6x4u, 2-ethyl-4-methyl-thiazole, fema no. 3680, pubchem9939, acmc-1buw7
PubChem CID 27440
IUPAC Name 2-ethyl-4-methyl-1,3-thiazole
SMILES CCC1=NC(=CS1)C

Specifications

Color Yellow
Boiling Point 67°C
Quantity 5 g
UN Number 1993
Formula Weight 127.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethyl-4-methylthiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.