accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CSH3CN

CAS RN 15679-13-7

IUPAC Name: 4-methyl-2-(propan-2-yl)-1,3-thiazole
Synonyms: 2-isopropyl-4-methyl-1,3-thiazole
Molecular Weight (g/mol): 141.23
Molecular Formula: C7H11NS
InChi Key: OFLXNHNYPQPQKW-UHFFFAOYSA-N
SMILES: CC(C)C1=NC(C)=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results