Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Isopropyl-4-methylthiazole 98.0+%, TCI America™
SDP

Catalog No. I04235G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I04235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I04235G Supplier TCI America Supplier No. I04235G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 15679-13-7
Molecular Formula C7H11NS
Molecular Weight (g/mol) 141.232
MDL Number MFCD00036561
InChI Key OFLXNHNYPQPQKW-UHFFFAOYSA-N
Synonym 2-isopropyl-4-methylthiazole, 4-methyl-2-1-methylethyl thiazole, 2-isopropyl-4-methyl thiazole, thiazole, 2-isopropyl-4-methyl, thiazole, 4-methyl-2-1-methylethyl, unii-nqf7jo3mup, tropical thiazole, 2-isopropyl-4-methyl-1,3-thiazole, fema no. 3555, nqf7jo3mup
PubChem CID 61808
IUPAC Name 4-methyl-2-propan-2-yl-1,3-thiazole
SMILES CC1=CSC(=N1)C(C)C

Specifications

Boiling Point 92°C
Quantity 5 g
Formula Weight 141.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Isopropyl-4-methylthiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.