CAS RN 15679-19-3
IUPAC Name:
2-ethoxy-1,3-thiazole
Synonyms:
2-ethoxy-1,3-thiazole
Molecular Weight (g/mol):
129.18
Molecular Formula:
C5H7NOS
InChi Key:
NDUWJHRKDYXRAD-UHFFFAOYSA-N
SMILES:
CCOC1=NC=CS1
1
–
1
of
1
results
1
2-Ethoxythiazole 98.0+%, TCI America™
CAS: 15679-19-3 Molecular Formula: C5H7NOS Molecular Weight (g/mol): 129.177 InChI Key: NDUWJHRKDYXRAD-UHFFFAOYSA-N PubChem CID: 61809 IUPAC Name: 2-ethoxy-1,3-thiazole SMILES: CCOC1=NC=CS1