accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1569-01-3

IUPAC Name: 1-propoxypropan-2-ol
Synonyms: 1-propoxy-2-propanol
Molecular Weight (g/mol): 118.18
Molecular Formula: C6H14O2
InChi Key: FENFUOGYJVOCRY-UHFFFAOYNA-N
SMILES: CCCOCC(C)O
Boiling Point 
  • (5)
  • (2)
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)