accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NSOOCH3HCl

CAS RN 156907-84-5

IUPAC Name: 3-(3-methanesulfonylphenyl)-1-propylpiperidine hydrochloride
Synonyms: 3-(3-methanesulfonylphenyl)-1-propylpiperidine hydrochloride
Molecular Weight (g/mol): 317.87
Molecular Formula: C15H24ClNO2S
InChi Key: LEMGVHZVBREXAD-UHFFFAOYNA-N
SMILES: Cl.CCCN1CCCC(C1)C1=CC(=CC=C1)S(C)(=O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results