Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ OSU 6162 hydrochloride
Description
Dopamine stabilizer; lacks high affinity for various neuroreceptors in vitro (Ki values are 447 and 1305 nM for D2 and D3 receptors respectively and > 1 μM for other targets). Exhibits high D2 receptor occupancy in vivo; highly active on dopamine synthesis and turnover. Induces stabilizing effects on psychomotor function in behavioral tests without inducing hypolocomotion or catalepsy.
Specifications
Specifications
| Quantity | 50 mg |
| Synonym | osu 6162 hydrochloride, osu6162 hydrochloride, 3s-3-3-methylsulfonyl phenyl-1-propylpiperidine hydrochloride, pnu-96391; pnu-0096391; osu-6162; s---3-methanesulfonyl-phenyl-1-propyl-piperidine hydrochloride; s,s-3-3-methylsulfonyl phenyl-1-propylpiperidine hydrochloride, 3s-3-3-methanesulfonylphenyl-1-propylpiperidine hydrochloride, unii-v5sbjbujw6, v5sbjbujw6, osu-6162 hydrochloride, osu6162 hydrochloride hplc, 3-3-methanesulfonyl-phenyl-1-propyl-piperidine hydrochloride |
| Formula Weight | 317.87 |
| Percent Purity | >98% |
| Chemical Name or Material | OSU 6162 hydrochloride |
Safety and Handling
| Recommended Storage | Store at +4°C |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?