accessibility menu, dialog, popup

UserName

Viewing profile as user:
COCOCOCOCOCOCOCOCOCOCOCOOsOsOs

CAS RN 15696-40-9

IUPAC Name: dodecakis(methanidylidyneoxidanium) triosmium
Synonyms: triosmium dodecacarbonyl
Molecular Weight (g/mol): 906.81
Molecular Formula: C12O12Os3
InChi Key: VUBLMKVEIPBYME-UHFFFAOYSA-N
SMILES: [Os].[Os].[Os].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)