accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 157026-18-1

IUPAC Name: 4-bromo-2-nitro-6-(trifluoromethyl)aniline
Synonyms: 4-bromo-2-nitro-6-(trifluoromethyl)aniline
Molecular Weight (g/mol): 285.02
Molecular Formula: C7H4BrF3N2O2
InChi Key: JWIAFKYOENFPNQ-UHFFFAOYSA-N
SMILES: NC1=C(C=C(Br)C=C1C(F)(F)F)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results