CAS RN 15707-34-3
IUPAC Name:
5-ethyl-2,3-dimethylpyrazine
Synonyms:
5-ethyl-2,3-dimethylpyrazine
Molecular Weight (g/mol):
136.2
Molecular Formula:
C8H12N2
InChi Key:
CIBKSMZEVHTQLG-UHFFFAOYSA-N
SMILES:
CCC1=CN=C(C)C(C)=N1
1
–
1
of
1
results
1
5-Ethyl-2,3-dimethylpyrazine 98.0+%, TCI America™
CAS: 15707-34-3 Molecular Formula: C8H12N2 Molecular Weight (g/mol): 136.20 MDL Number: MFCD04038059 InChI Key: CIBKSMZEVHTQLG-UHFFFAOYSA-N PubChem CID: 27460 IUPAC Name: 5-ethyl-2,3-dimethylpyrazine SMILES: CCC1=CN=C(C)C(C)=N1