Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Ethyl-2,3-dimethylpyrazine 98.0+%, TCI America™
SDP

Catalog No. E082825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E082825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E082825G Supplier TCI America Supplier No. E082825G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 15707-34-3
Molecular Formula C8H12N2
Molecular Weight (g/mol) 136.20
MDL Number MFCD04038059
InChI Key CIBKSMZEVHTQLG-UHFFFAOYSA-N
PubChem CID 27460
IUPAC Name 5-ethyl-2,3-dimethylpyrazine
SMILES CCC1=CN=C(C)C(C)=N1

Specifications

Color Yellow
Boiling Point 76°C
Quantity 25 g
Formula Weight 136.20
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 5-Ethyl-2,3-dimethylpyrazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.