accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClClOHNOCl

CAS RN 1571-13-7

IUPAC Name: 4,5,6,7-tetrachloro-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms: 4,5,6,7-tetrachloro-2H-isoindole-1,3-dione
Molecular Weight (g/mol): 284.9
Molecular Formula: C8HCl4NO2
InChi Key: LPUUYZVKCMCHLO-UHFFFAOYSA-N
SMILES: ClC1=C2C(=O)NC(=O)C2=C(Cl)C(Cl)=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results