accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1571-75-1

IUPAC Name: 4-[1-(4-hydroxyphenyl)-1-phenylethyl]phenol
Synonyms: 4-[1-(4-hydroxyphenyl)-1-phenylethyl]phenol
Molecular Weight (g/mol): 290.36
Molecular Formula: C20H18O2
InChi Key: VOWWYDCFAISREI-UHFFFAOYSA-N
SMILES: CC(C1=CC=CC=C1)(C1=CC=C(O)C=C1)C1=CC=C(O)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)