CAS RN 15715-41-0
IUPAC Name:
diethyl methylphosphonite
Synonyms:
diethyl methylphosphonite
Molecular Weight (g/mol):
136.13
Molecular Formula:
C5H13O2P
InChi Key:
NSSMTQDEWVTEKN-UHFFFAOYSA-N
SMILES:
CCOP(C)OCC
1
–
1
of
1
results
1
Sigma Aldrich Diethyl Methylphosphonite
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.